Invitrogen
Manufacturer Code:PA123323
Catalog # PIPA123323
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein encoding the full-length p37 subunit ofhuman RFC purified from E. coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Polyclonal Antibody
For Research Use Only
Protein Aliases: A1 37 kDa subunit; Activator 1 37 kDa subunit; Activator 1 subunit 4; replication factor C (activator 1) 4, 37kDa; Replication factor C 37 kDa subunit; Replication factor C subunit 4; RF-C 37 kDa subunit; RFC 37 kDa subunit; RFC p37
Gene Aliases: A1; RFC37; RFC4
UniProt ID: (Human) P35249
Entrez Gene ID: (Human) 5984
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.