Novus Biologicals
Manufacturer Code:NB100403
Catalog # NB100403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 23 amino acid synthetic peptide which represented a portion of the human Replication Factor C 4 37 kDa subunit encoded within exon 2 (locusLink ID 5984). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C. Do not freeze. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1 37 kDa subunit; A1 A1 37 kDa subunit Activator 1 37 kDa subunit Activator 1 subunit 4 MGC27291 replication factor C (activator 1) 4 37kDa Replication factor C 37 kDa subunit replication factor C subunit 4 RFC 37 kDa subunit RF-C 37 kDa subunit RFC37replication factor C (activator 1) 4 (37kD); Activator 1 37 kDa subunit; Activator 1 subunit 4; replication factor C (activator 1) 4, 37kDa; Replication factor C 37 kDa subunit; Replication factor C subunit 4; RF-C 37 kDa subunit; RFC 37 kDa subunit
Gene Aliases: A1; RFC37; RFC4
UniProt ID: (Human) P35249
Entrez Gene ID: (Human) 5984
Molecular Function:
DNA binding protein
DNA-directed DNA polymerase
nucleic acid binding
nucleotidyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.