Novus Biologicals
Manufacturer Code:NBP203934
Catalog # NBP203934
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4C11 |
Immunogen | Human recombinant protein fragment corresponding to amino acids 1-234 of human RFC2 (NP_002905) produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: A1 40 kDa subunit; A1 A1 40 kDa subunit Activator 1 40 kDa subunit Activator 1 subunit 2 MGC3665 replication factor C (activator 1) 2 40kDa replication factor C 2 Replication factor C 40 kDa subunit replication factor C subunit 2 RF-C 40 kDa subunit RFC40replication factor C (activator 1) 2 (40kD); Activator 1 40 kDa subunit; Activator 1 subunit 2; replication factor C (activator 1) 2, 40kDa; Replication factor C 40 kDa subunit; Replication factor C subunit 2; RF-C 40 kDa subunit
Gene Aliases: RFC2; RFC40
UniProt ID: (Human) P35250
Entrez Gene ID: (Human) 5982
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.