Invitrogen
Manufacturer Code:PA556169
Catalog # PIPA556169
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RFC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: A1 40 kDa subunit; A1 RFC40 A1 40 kDa subunit RF-C 40 kDa subunit activator 1 40 kDa subunit activator 1 subunit 2 replication factor C 2 replication factor C subunit 2; Activator 1 40 kDa subunit; Activator 1 subunit 2; Activator 1 subunit C2; replication factor C (activator 1) 2, 40kDa; Replication factor C 40 kDa subunit; Replication factor C subunit 2; replication factor C, 40 kDa; RF-C 40 kDa subunit; RFC40
Gene Aliases: 2610008M13Rik; 40kDa; AI326953; Recc2; RFC2; RFC40
UniProt ID: (Human) P35250, (Mouse) Q9WUK4, (Rat) Q641W4
Entrez Gene ID: (Human) 5982, (Mouse) 19718, (Rat) 116468
Molecular Function: DNA binding protein DNA-directed DNA polymerase nucleic acid binding nucleotidyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.