Abnova
Manufacturer Code:PAB11364
Catalog # 89-115-046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Baker's yeast |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids 115-129 of Saccharomyces cerevisiae RFA2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RFA2.
For Research Use Only
Protein Aliases: BUF1; DNA-binding protein BUF1; Replication factor A protein 2; Replication protein A 36 kDa subunit; RF-A protein 2; Rfa2p; RPA2
Gene Aliases: BUF1; N0368; RFA2; RPA2; RPA32; YNL312W
UniProt ID: (Baker's yeast) P26754
Entrez Gene ID: (Baker's yeast) 855404
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.