Invitrogen
Manufacturer Code:PA5100870
Catalog # PIPA5100870
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human REV1 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1110027I23Rik AIBP80 Alpha integrin-binding protein 80 AU022044 DNA repair protein REV1 REV1 REV1 homolog REV1 homolog (S. cerevisiae) REV1- like REV1 DNA directed polymerase REV1 polymerase (DNA directed) REV1L Rev1-like terminal deoxycytidyl transferase; AIBP80; Alpha integrin-binding protein 80; DNA repair protein REV1; REV1 homolog; REV1, polymerase (DNA directed); REV1- like; Rev1-like terminal deoxycytidyl transferase
Gene Aliases: 1110027I23Rik; AIBP80; AU022044; REV1; REV1L
UniProt ID: (Human) Q9UBZ9, (Mouse) Q920Q2
Entrez Gene ID: (Human) 51455, (Mouse) 56210
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.