Invitrogen
Manufacturer Code:MA535361
Catalog # PIMA535361
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0755 |
Immunogen | A synthesized peptide derived from human REST / NRSF |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 2610008J04Rik AA407358 Neural-restrictive silencer factor NRSF RE1-silencing transcription factor Rest REST4 WT6 X2 box repressor XBR; Neural-restrictive silencer factor; RE1-silencing transcription factor; zinc finger transcription factor REST protein
Gene Aliases: 2610008J04Rik; AA407358; NRSF; Rest; REST4
UniProt ID: (Mouse) Q8VIG1, (Rat) O54963
Entrez Gene ID: (Mouse) 19712, (Rat) 83618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.