Invitrogen
Manufacturer Code:PA5102923
Catalog # PIPA5102923
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human REST corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2610008J04Rik AA407358 Neural-restrictive silencer factor neuron restrictive silencer factor neuron-restrictive silencer factor NRSF RE1 silencing transcription factor RE1-silencing transcription factor RE1-silencing transcription factor variant E1a/E2/E3/E4c RE1-silencing transcription factor variant E1a/E2/E3/E5 RE1-silencing transcription factor variant E1a/E2/E3/N3a/E4i RE1-silencing transcription factor variant E1a/E2/E3/N3c/E4 RE1-silencing transcription factor variant E1a/E2/E4 RE1-silencing transcription factor variant E1a/E2/E5 RE1-silencing transcription factor variant E1a/E2a/E2k RE1-silencing transcription factor variant E1a/E2d/E4g RE1-silencing transcription factor variant E1a/E2e/E4h RE1-silencing transcription factor variant E1a/E2f/E4e RE1-silencing transcription factor variant E1a/E2k/E2i/E3/E4j RE1-silencing transcription factor variant E1b/E2/E3/E5 RE1-silencing transcription factor variant E1b/E2/E3/N3b/E4i RE1-silencing transcription factor variant E1b/E2/E3/N3c/E4 RE1-silencing transcription factor variant E1b/E2a/E2k RE1-silencing transcription factor variant E1b/E2c/E2j/E3/E4 RE1-silencing transcription factor variant E1b/E2e/E4h RE1-silencing transcription factor variant E1c/E2/E3/E5 RE1-silencing transcription factor variant E1c/E2a/E2k RE1-silencing transcription factor variant E1c/E2g/E3/E4 repressor binding to the X2 box REST REST4 WT6 X2 box repressor XBR zinc finger transcription factor REST protein; Neural-restrictive silencer factor; neuron restrictive silencer factor; neuron-restrictive silencer factor; RE1-silencing transcription factor; RE1-silencing transcription factor variant E1a/E2/E3/E4c; RE1-silencing transcription factor variant E1a/E2/E3/E5; RE1-silencing transcription factor variant E1a/E2/E3/N3a/E4i; RE1-silencing transcription factor variant E1a/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1a/E2/E4; RE1-silencing transcription factor variant E1a/E2/E5; RE1-silencing transcription factor variant E1a/E2a/E2k; RE1-silencing transcription factor variant E1a/E2d/E4g; RE1-silencing transcription factor variant E1a/E2e/E4h; RE1-silencing transcription factor variant E1a/E2f/E4e; RE1-silencing transcription factor variant E1a/E2k/E2i/E3/E4j; RE1-silencing transcription factor variant E1b/E2/E3/E5; RE1-silencing transcription factor variant E1b/E2/E3/N3b/E4i; RE1-silencing transcription factor variant E1b/E2/E3/N3c/E4; RE1-silencing transcription factor variant E1b/E2a/E2k; RE1-silencing transcription factor variant E1b/E2c/E2j/E3/E4; RE1-silencing transcription factor variant E1b/E2e/E4h; RE1-silencing transcription factor variant E1c/E2/E3/E5; RE1-silencing transcription factor variant E1c/E2a/E2k; RE1-silencing transcription factor variant E1c/E2g/E3/E4; repressor binding to the X2 box; X2 box repressor; zinc finger transcription factor REST protein
Gene Aliases: NRSF; REST; WT6; XBR
UniProt ID: (Human) Q13127, (Rat) O54963
Entrez Gene ID: (Human) 5978, (Rat) 83618
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.