Invitrogen
Manufacturer Code:PA540586
Catalog # PIPA540586
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RERE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: arginine-glutamic acid dipeptide (RE) repeats; Arginine-glutamic acid dipeptide (RE) repeats ARG ARP ATN1L DNB1 FLJ38775 KIAA0458; Arginine-glutamic acid dipeptide repeats protein; atrophin 2; atrophin-1 like protein; atrophin-1 related protein; Atrophin-1-like protein; Atrophin-1-related protein; Atrophin-2
Gene Aliases: 1110033A15Rik; AI414665; ARG; ARP; ATN1L; Atr2; AW742570; DNB1; eye<m03Jus>; eyes3; KIAA0458; mKIAA0458; NEDBEH; RERE
UniProt ID: (Human) Q9P2R6, (Rat) Q62901, (Mouse) Q80TZ9
Entrez Gene ID: (Horse) 100051942, (Human) 473, (Rabbit) 100358540, (Guinea pig) 100721896, (Rat) 116665, (Mouse) 68703
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.