Novus Biologicals
Manufacturer Code:NBP2856250.1ML
Catalog # NB033554
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of human RERE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ARGARPKIAA0458arginine-glutamic acid dipeptide repeats protein arginine-glutamic acid dipeptide (RE) repeats ATN1L atrophin 2 atrophin-1 like protein atrophin-1 related protein Atrophin-1-like protein Atrophin-1-related protein DNB1 FLJ38775; arginine-glutamic acid dipeptide (RE) repeats; Arginine-glutamic acid dipeptide repeats protein; atrophin 2; atrophin-1 like protein; atrophin-1 related protein; Atrophin-1-like protein; Atrophin-1-related protein
Gene Aliases: ARG; ARP; ATN1L; DNB1; KIAA0458; NEDBEH; RERE
UniProt ID: (Human) Q9P2R6
Entrez Gene ID: (Human) 473
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.