Abnova
Manufacturer Code:H00029803A01
Catalog # 89-106-789
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 1 a.a. &bsim 110 a.a) partial recombinant protein with GST tag.,REPIN1 (NP_037532 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant REPIN1.
For Research Use Only
Protein Aliases: 60 kDa origin-specific DNA-binding protein; 60 kDa replication initiation region protein; AP4; ATT-binding protein; DHFR oribeta-binding protein RIP60; H_DJ0584D14.12; replication initiation region protein (60kD); Replication initiator 1; RIP60; Zfp464; Zinc finger protein 464; zinc finger protein 464 (RIP60); zinc finger protein AP4; ZNF464
Gene Aliases: AP4; REPIN1; RIP60; Zfp464; ZNF464
UniProt ID: (Human) Q9BWE0
Entrez Gene ID: (Human) 29803
Molecular Function: KRAB box transcription factor transcription factor zinc finger transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.