Novus Biologicals
Manufacturer Code:NBP216905
Catalog # NBP216905
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein encompassing a sequence within the N-terminus region of human RENT1. The exact sequence is proprietary. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ATP-dependent helicase RENT1; ATP-dependent helicase RENT1 EC 3.6.1 EC 3.6.4.- FLJ43809 HUPF1 KIAA0221FLJ46894 Nonsense mRNA reducing factor 1 NORF1delta helicase pNORF1 regulator of nonsense transcripts 1 RENT1UP Frameshift 1 UPF1 regulator of nonsense transcripts homolog (yeast) up-frameshift mutation 1 homolog Up-frameshift suppressor 1 homolog yeast Upf1p homolog; delta helicase; hUpf1; Nonsense mRNA reducing factor 1; NORF1; Regulator of nonsense transcripts 1; smg-2 homolog, nonsense mediated mRNA decay factor; up-frameshift mutation 1 homolog; Up-frameshift suppressor 1 homolog; UPF1 regulator of nonsense transcripts homolog; yeast Upf1p homolog
Gene Aliases: HUPF1; KIAA0221; NORF1; pNORF1; RENT1; smg-2; UPF1
UniProt ID: (Human) Q92900
Entrez Gene ID: (Human) 5976
Molecular Function:
DNA binding protein
DNA helicase
RNA binding protein
RNA helicase
endoribonuclease
helicase
hydrolase
nuclease
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.