Invitrogen
Manufacturer Code:PA551387
Catalog # PIPA551387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RENBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGE; GlcNAc 2-epimerase; N-acetyl-D-glucosamine 2-epimerase; N-acylglucosamine 2-epimerase; Renin-binding protein; RnBP
Gene Aliases: Age; RBP; RENBP; RNBP
UniProt ID: (Human) P51606, (Mouse) P82343, (Rat) P51607
Entrez Gene ID: (Human) 5973, (Mouse) 19703, (Rat) 81759
Molecular Function: epimerase/racemase isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.