Novus Biologicals
Manufacturer Code:NBP288139
Catalog # NB035761
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the N-terminal region of RENBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AGE; AGE EC 5.1.3.8 GlcNAc 2-epimerase N-acetyl-D-glucosamine 2-epimerase N-acylglucosamine 2-epimerase RBP renin binding protein Renin-binding protein RNBP; GlcNAc 2-epimerase; N-acetyl-D-glucosamine 2-epimerase; N-acylglucosamine 2-epimerase; Renin-binding protein; RnBP
Gene Aliases: RBP; RENBP; RNBP
UniProt ID: (Human) P51606
Entrez Gene ID: (Human) 5973
Molecular Function:
epimerase/racemase
isomerase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.