Proteintech
Manufacturer Code:150031AP150UL
Catalog # 50-172-9901
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RENALASE Fusion Protein Ag13061 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: alpha-NAD(P)H oxidase/anomerase; C10orf59 MAO C Monoamine oxidase C RENALASE RNLS; MAO-C; mMAO-C; Monoamine oxidase-C; Renalase; renalase, FAD-dependent amine oxidase; rnls
Gene Aliases: 6530404N21Rik; AI452315; AW060440; C10orf59; RENALASE; RGD1309804; RNLS
UniProt ID: (Human) Q5VYX0, (Rat) Q5U2W9
Entrez Gene ID: (Human) 55328, (Rat) 361751, (Mouse) 67795
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.