Invitrogen
Manufacturer Code:MA530810
Catalog # PIMA530810
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 5 |
Immunogen | Recombit Human REG4 Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Gastrointestinal secretory protein; GISP RELP Gastrointestinal secretory protein GISPREG-4 Reg IV regenerating gene type IV regenerating islet-derived family member 4 regenerating islet-derived protein 4 Regenerating islet-derived protein IV REG-IV RELPREG-like protein; Reg IV; REG-4; REG-like protein; regenerating gene type IV; regenerating islet-derived family, member 4; Regenerating islet-derived protein 4; Regenerating islet-derived protein IV
Gene Aliases: GISP; REG-IV; REG4; RELP
UniProt ID: (Human) Q9BYZ8
Entrez Gene ID: (Human) 83998
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.