Invitrogen
Manufacturer Code:PA575710
Catalog # PIPA575710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to residues in Human REG4 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Gastrointestinal secretory protein; Reg IV; REG-4; REG-like protein; regenerating gene type IV; regenerating islet-derived family, member 4; Regenerating islet-derived protein 4; Regenerating islet-derived protein 4 REG-4 Gastrointestinal secretory protein REG-like protein Regenerating islet-derived protein IV Reg IV REG4 GISP RELP ITGA5 FNRA; Regenerating islet-derived protein IV
Gene Aliases: 2010002L15Rik; GISP; REG-IV; REG4; RELP
UniProt ID: (Human) Q9BYZ8, (Mouse) Q9D8G5, (Rat) Q68AX7
Entrez Gene ID: (Human) 83998, (Mouse) 67709, (Rat) 445583
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.