Novus Biologicals
Manufacturer Code:NBP176255
Catalog # NBP176255
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Antibody was raised against a 17 amino acid synthetic peptide near the center of human REEP4. The immunogen is located within amino acids 160 - 210 of REEP4. Amino Acid Squence: AQRWQPGSRGEKVTGGY |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C8orf20chromosome 8 open reading frame 20 FLJ22246 FLJ22277 PP432 receptor accessory protein 4 receptor expression enhancing protein 4 receptor expression-enhancing protein 4; receptor expression enhancing protein 4; Receptor expression-enhancing protein 4
Gene Aliases: C8orf20; PP432; REEP4; Yip2c
UniProt ID: (Human) Q9H6H4
Entrez Gene ID: (Human) 80346
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.