Proteintech
Manufacturer Code:266501AP150UL
Catalog # 50-173-5413
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | REEP4 Fusion Protein Ag24569 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C8orf20 PP432 receptor accessory protein 4 REEP4; receptor expression enhancing protein 4; Receptor expression-enhancing protein 4
Gene Aliases: 2700029E10Rik; C8orf20; PP432; REEP4; RGD1306561; Yip2c
UniProt ID: (Human) Q9H6H4, (Rat) Q4QQW1, (Mouse) Q8K072
Entrez Gene ID: (Human) 80346, (Rat) 306014, (Mouse) 72549
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.