OriGene
Manufacturer Code:CF504322
Catalog # 50-166-2805
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2B7 |
Immunogen | Full length human recombit protein of human REEP2 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: C5orf19 SGC32445 SPG72 Yip2d; Receptor expression-enhancing protein 2
Gene Aliases: C5orf19; REEP2; SGC32445; SPG72; Yip2d
UniProt ID: (Human) Q9BRK0, (Mouse) Q8VCD6
Entrez Gene ID: (Dog) 100683708, (Human) 51308, (Rat) 682105, (Mouse) 225362
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.