Novus Biologicals
Manufacturer Code:NBP232017
Catalog # NBP232017
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: SPGSLLDTIEDLGDDPALSLRSSTNPADSRTEASEDDMGDKAPKRAKPIKKAPKAEPLASKTLKTRPKKKTSGG |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C5orf19receptor expression-enhancing protein 2 chromosome 5 open reading frame 19 receptor accessory protein 2 SGC32445receptor expression enhancing protein 2; Receptor expression-enhancing protein 2
Gene Aliases: C5orf19; REEP2; SGC32445; SPG72; Yip2d
UniProt ID: (Human) Q9BRK0
Entrez Gene ID: (Human) 51308
Molecular Function:
receptor
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.