Proteintech
Manufacturer Code:179881AP150UL
Catalog # 50-173-1920
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | REEP1 Fusion Protein Ag12418 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C2orf23 receptor accessory protein 1 REEP1 SPG31; Receptor expression-enhancing protein 1; Spastic paraplegia 31 protein
Gene Aliases: C2orf23; D6Ertd253e; HMN5B; REEP1; RGD1305230; SPG31; Yip2a
UniProt ID: (Human) Q9H902, (Mouse) Q8BGH4
Entrez Gene ID: (Human) 65055, (Mouse) 52250, (Rat) 362384
Molecular Function: receptor transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.