Novus Biologicals
Manufacturer Code:NBP294794
Catalog # NB024101
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 180-380 of human ADARB1 (NP_001103.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: ADAR2a ADAR2ADAR2g ADAR2a-L1 ADAR2a-L2 ADAR2a-L3 ADAR2b ADAR2c adenosine deaminase RNA-specific B1 double-stranded RNA-specific editase 1 DRABA2 DRADA2hRED1 EC 3.5 human dsRNA adenosine deaminase DRADA2 human dsRNA adenosine deaminase DRADA2b EC 3.510adenosine deaminase RNA-specific B1 (homolog of rat RED1) RED1 homolog RED1ADAR2d RNA editase RNA editing deaminase 1 RNA-editing deaminase 1 RNA-editing enzyme 1 RNA-specific B1 (RED1 homolog rat); adenosine deaminase, RNA-specific, B1 (homolog of rat RED1); adenosine deaminase, RNA-specific, B1 (RED1 homolog rat); Double-stranded RNA-specific editase 1; dsRNA adenosine deaminase; dsRNA adenosine deaminase DRADA2; RED1 homolog; RNA editase; RNA editing deaminase 1; RNA-editing deaminase 1; RNA-editing enzyme 1
Gene Aliases: ADAR2; ADARB1; DRABA2; DRADA2; RED1
UniProt ID: (Human) P78563
Entrez Gene ID: (Human) 104
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.