Invitrogen
Manufacturer Code:PA558423
Catalog # PIPA558423
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human RDH16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: hRDH-E; Human epidermal retinol dehydrogenase; Microsomal NAD(+)-dependent retinol dehydrogenase 4; microsomal NAD+-dependent retinol dehydrogenase 4; Retinol dehydrogenase 16; retinol dehydrogenase 16 (all-trans and 13-cis); RoDH-4; RODH-4 SDR9C8 microsomal NAD+-dependent retinol dehydrogenase 4 retinol dehydrogenase 16 retinol dehydrogenase 16 (all-trans and 13-cis) short chain dehydrogenase/reductase family 9C member 8 sterol/retinol dehydrogenase; Short chain dehydrogenase/reductase family 9C member 8; short chain dehydrogenase/reductase family 9C, member 8; Sterol/retinol dehydrogenase
Gene Aliases: RDH16; RODH-4; RODH4; SDR9C8
UniProt ID: (Human) O75452
Entrez Gene ID: (Human) 8608
Molecular Function: dehydrogenase oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.