Invitrogen
Manufacturer Code:PA542115
Catalog # PIPA542115
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RDH16 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 29 kDa protein; Cis-retinol androgen dehydrogenase 1; cis-retinol/androgen dehydrogenase 1; hRDH-E; Human epidermal retinol dehydrogenase; Microsomal NAD(+)-dependent retinol dehydrogenase 4; microsomal NAD+-dependent retinol dehydrogenase 4; Retinol dehydrogenase 16; retinol dehydrogenase 16 (all-trans and 13-cis); retinol dehydrogenase 16 (all-trans); Retinol dehydrogenase 16 (all-trans) RODH-4 SDR9C8; Retinol dehydrogenase 2; retinol dehydrogenase 6; Retinol dehydrogenase type 1; retinol dehydrogenase type 2; retinol dehydrogenase type 6; Retinol dehydrogenase type II; retinol dehydrogenase type II (RODH II); RODH II; RoDH-4; Short chain dehydrogenase/reductase family 9C member 8; short chain dehydrogenase/reductase family 9C, member 8; Sterol/retinol dehydrogenase
Gene Aliases: CRAD; CRAD1; Rdh1; RDH16; Rdh2; Rdh6; RODH-4; Rodh1; RODH4; RoDHII; SDR9C8
UniProt ID: (Human) O75452, (Mouse) O54909, (Rat) P50170
Entrez Gene ID: (Bovine) 519940, (Dog) 474409, (Human) 8608, (Pig) 100511633, (Mouse) 19683, (Rat) 299511
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.