Proteintech
Manufacturer Code:162831AP150UL
Catalog # 50-173-0851
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RDH14 Fusion Protein Ag9386 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Alcohol dehydrogenase PAN2; Alcohol dehydrogenase PAN2 PAN2 RDH14 Retinol dehydrogenase 14 SDR7C4; alcohol dehydrogenase PAN2; expressed sequence AW215439; pancreas protein 2; Retinol dehydrogenase 14; retinol dehydrogenase 14 (all-trans and 9-cis); retinol dehydrogenase 14 (all-trans and 9-cis)9; Short chain dehydrogenase/reductase family 7C member 4; short chain dehydrogenase/reductase family 7C, member 4
Gene Aliases: 3110030G19Rik; AW215439; PAN2; RDH14; RGD1565196; SDR7C4; UNQ529/PRO1072
UniProt ID: (Human) Q9HBH5, (Mouse) Q9ERI6
Entrez Gene ID: (Human) 57665, (Mouse) 105014, (Rat) 500629
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.