Proteintech
Manufacturer Code:160671AP150UL
Catalog # 50-173-0688
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RDH13 Fusion Protein Ag8944 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RDH13 Retinol dehydrogenase 13 SDR7C3 UNQ736/PRO1430; Retinol dehydrogenase 13; retinol dehydrogenase 13 (all-trans and 9-cis); Short chain dehydrogenase/reductase family 7C member 3; short chain dehydrogenase/reductase family 7C, member 3
Gene Aliases: 8430425D21Rik; PSEC0082; RDH13; SDR7C3; UNQ736/PRO1430
UniProt ID: (Human) Q8NBN7, (Mouse) Q8CEE7
Entrez Gene ID: (Human) 112724, (Rat) 361504, (Mouse) 108841
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.