Novus Biologicals
Manufacturer Code:NBP273843AF350
Catalog # NP273843A30
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B4 |
Immunogen | Full length recombinant protein of human RDH11 (NP_057110) produced in HEK293T cells. |
Conjugate | Alexa Fluor 350 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Androgen-regulated short-chain dehydrogenase/reductase 1; Androgen-regulated short-chain dehydrogenase/reductase 1 ARSDR1CGI82 EC 1.1.1.300 HCBP12 HCV core-binding protein HCBP12 MDT1 prostate short-chain dehydrogenase reductase 1 Prostate short-chain dehydrogenase/reductase 1 PSDR1FLJ32633 RALR1 Retinal reductase 1 retinol dehydrogenase 11 retinol dehydrogenase 11 (all-trans and 9-cis) retinol dehydrogenase 11 (all-trans/9-cis/11-cis) SCALD SDR7C1 short chain dehydrogenase/reductase family 7C member 1; HCV core-binding protein HCBP12; prostate short-chain dehydrogenase reductase 1; Prostate short-chain dehydrogenase/reductase 1; RalR1; Retinal reductase 1; Retinol dehydrogenase 11; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1
Gene Aliases: ARSDR1; CGI-82; CGI82; HCBP12; MDT1; PSDR1; RALR1; RDH11; RDJCSS; SCALD; SDR7C1
UniProt ID: (Human) Q8TC12
Entrez Gene ID: (Human) 51109
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.