Abnova
Manufacturer Code:PAB5925
Catalog # 89-132-012
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of mouse Rdh11. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Rdh11.
For Research Use Only
Protein Aliases: 2610319N22Rik; AI428145; all-trans and 9-cis; Androgen-regulated short-chain dehydrogenase/reductase 1; Arsdr1; AU045252; C85936; cell line MC/9.IL4 derived transcript 1; Cell line MC/9.IL4-derived protein 1; CGI-82; HCBP12; M42C60; Mdt1; Prostate short-chain dehydrogenase/reductase 1; Psdr1; RalR1; Retinal reductase 1; Retinol dehydrogenase 11; SCALD; Short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; Ube-1c; UBE-1c1; ubiquitously expressing
Gene Aliases: 2610319N22Rik; AI428145; Arsdr1; AU045252; C85936; CGI-82; HCBP12; M42C60; Mdt1; Psdr1; Rdh11; SCALD; Ube-1c; UBE-1c1
UniProt ID: (Mouse) Q9QYF1
Entrez Gene ID: (Mouse) 17252
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.