Proteintech
Manufacturer Code:110771AP150UL
Catalog # 50-172-6855
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RDH11 Fusion Protein Ag1485 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: all-trans and 9-cis; Androgen-regulated short-chain dehydrogenase/reductase 1; ARSDR1 CGI 82 HCBP12 MDT1 PSDR1 RALR1 RDH11 Retinal reductase 1 Retinol dehydrogenase 11 SCALD SDR7C1; cell line MC/9.IL4 derived transcript 1; Cell line MC/9.IL4-derived protein 1; HCV core-binding protein HCBP12; M42C60; prostate short-chain dehydrogenase reductase 1; Prostate short-chain dehydrogenase/reductase 1; RalR1; Retinal reductase 1; Retinol dehydrogenase 11; SCALD; Short chain dehydrogenase/reductase family 7C member 1; short chain dehydrogenase/reductase family 7C, member 1; Short-chain aldehyde dehydrogenase; short-chain dehydrogenase/reductase 1; ubiquitously expressing
Gene Aliases: 2610319N22Rik; AI428145; ARSDR1; AU045252; C85936; CGI-82; CGI82; HCBP12; M42C60; MDT1; PSDR1; RALR1; RDH11; RDJCSS; SCALD; SDR7C1; Ube-1c; UBE-1c1
UniProt ID: (Human) Q8TC12, (Mouse) Q9QYF1
Entrez Gene ID: (Human) 51109, (Mouse) 17252, (Rat) 362757
Molecular Function:
dehydrogenase
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.