Invitrogen
Manufacturer Code:PA564918
Catalog # PIPA564918
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human RDBP |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: major histocompatibility complex gene RD; Negative elongation factor E; negative elongation factor polypeptide E; NELF-E; nuclear protein; RD RNA binding protein; RD RNA binding protein D6S45 NELF-E RD RDP RDBP; RD RNA-binding protein; RNA-binding protein RD
Gene Aliases: D6S45; NELF-E; NELFE; RD; RDBP; RDP
UniProt ID: (Human) P18615
Entrez Gene ID: (Human) 7936
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.