Abnova
Manufacturer Code:MAB11927
Catalog # 89-110-064
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S72-8 |
Immunogen | Recombinant fusion protein corresponding to amino acids of 109-293,including ELM2 and SANT domains,of human RCOR1. |
Conjugate | ATTO 390 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against partial recombinant RCOR1.
For Research Use Only
Protein Aliases: COREST; KIAA0071; Protein CoREST; RCOR; REST corepressor 1
Gene Aliases: COREST; KIAA0071; RCOR; RCOR1
UniProt ID: (Human) Q9UKL0
Entrez Gene ID: (Human) 23186
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.