Invitrogen
Manufacturer Code:PA541564
Catalog # PIPA541564
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human RCOR1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: corest; Protein CoREST; REST corepressor 1; REST corepressor 1 COREST KIAA0071 RCOR
Gene Aliases: 5730409O11; 6720480E22Rik; AU042633; COREST; D12Wsu95e; im:6904264; KIAA0071; mKIAA0071; RCOR; RCOR1; Rocr1; si:dkey-46n18.7; zgc:158607
UniProt ID: (Human) Q9UKL0, (Mouse) Q8CFE3
Entrez Gene ID: (Human) 23186, (Mouse) 217864, (Zebrafish) 569294
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.