Abnova
Manufacturer Code:PAB15801
Catalog # 89-117-864
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant GST fusion protein corresponding to 273 amino acids of mouse Rcor1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against partial recombinant Rcor1.
For Research Use Only
Protein Aliases: 5730409O11; 6720480E22Rik; AU042633; D12Wsu95e; mKIAA0071; Protein CoREST; REST corepressor 1
Gene Aliases: 5730409O11; 6720480E22Rik; AU042633; D12Wsu95e; Kiaa0071; mKIAA0071; Rcor1; Rocr1
UniProt ID: (Mouse) Q8CFE3
Entrez Gene ID: (Mouse) 217864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.