Invitrogen
Manufacturer Code:PA576587
Catalog # PIPA576587
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein of human RCL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: RNA 3'-terminal phosphate cyclase-like protein; RNA cyclase homolog; RNA terminal phosphate cyclase-like 1; RNAC RPCL1
Gene Aliases: 2310040A02Rik; Ab1-353; Ac1292; AI789745; C76567; HSPC338; RCL1; RNAC; RPC2; RPCL1; RTC2
UniProt ID: (Human) Q9Y2P8, (Mouse) Q9JJT0
Entrez Gene ID: (Human) 10171, (Mouse) 59028, (Rat) 309301
Molecular Function:
RNA binding protein
cyclase
lyase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.