Invitrogen
Manufacturer Code:PA544747
Catalog # PIPA544747
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human RCC2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Protein RCC2; Protein RCC2 homolog; RCC1-like protein TD-60; regulator of chromosome condensation 2; Telophase disk protein of 60 kDa
Gene Aliases: 2610510H01Rik; 2610529N02Rik; AA536646; AA675016; KIAA1470; mKIAA1470; RCC2; RGD1309986; TD-60; TD60; wu:fb36a03; zgc:77115
UniProt ID: (Human) Q8IVL9, (Mouse) Q8BK67, (Zebrafish) Q6NYE2
Entrez Gene ID: (Bovine) 509120, (Horse) 100052965, (Dog) 487412, (Human) 55920, (Rabbit) 100351463, (Guinea pig) 100734723, (Mouse) 108911, (Rat) 298594, (Zebrafish) 406455
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.