Novus Biologicals
Manufacturer Code:NBP297639IR
Catalog # NB045910
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human RCC2 fragment. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: DKFZp762N0610 KIAA1470 protein RCC2 RCC1-like protein TD-60 regulator of chromosome condensation 2 TD60 TD-60 Telophase disk protein of 60 kDa; Protein RCC2; RCC1-like protein TD-60; Telophase disk protein of 60 kDa
Gene Aliases: KIAA1470; RCC2; TD-60; TD60
UniProt ID: (Human) Q8IVL9
Entrez Gene ID: (Human) 55920
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.