Novus Biologicals
Manufacturer Code:NBP197543JF549
Catalog # NBP97543549
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3D11 |
Immunogen | Recombinant human RanGEF. |
Conjugate | Janelia Fluor 549 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Cell cycle regulatory protein; Cell cycle regulatory protein CHC1RCC1-I chromosome condensation 1 Chromosome condensation protein 1 guanine nucleotide-releasing protein regulator of chromosome condensation regulator of chromosome condensation 1 SNHG3-RCC1 SNHG3-RCC1 readthrough transcript; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754
Entrez Gene ID: (Human) 1104
Molecular Function: DNA binding protein G-protein modulator chromatin/chromatin-binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.