Invitrogen
Manufacturer Code:PA570527
Catalog # PIPA570527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human RCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle regulatory protein; CHC1 RCC1-I SNHG3-RCC1 SNHG3-RCC1 readthrough transcript cell cycle regulatory protein guanine nucleotide-releasing protein regulator of chromosome condensation; chromosome condensation 1; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: 4931417M11Rik; AI326872; CHC1; fj77f08; RCC1; RCC1-I; SNHG3-RCC1; wu:fj77f08; zgc:65796; zgc:77114
UniProt ID: (Human) P18754, (Mouse) Q8VE37
Entrez Gene ID: (Dog) 487332, (Human) 1104, (Horse) 100070733, (Rabbit) 100352855, (Bovine) 534142, (Mouse) 100088, (Guinea pig) 100719046, (Rat) 682908, (Zebrafish) 406457
Molecular Function:
DNA binding protein
G-protein modulator
chromatin/chromatin-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.