Invitrogen
Manufacturer Code:PA528063
Catalog # PIPA528063
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 2 and 421 of Human RCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle regulatory protein; CHC1 RCC1-I SNHG3-RCC1 SNHG3-RCC1 readthrough transcript cell cycle regulatory protein guanine nucleotide-releasing protein regulator of chromosome condensation; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: 4931417M11Rik; AI326872; CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754, (Mouse) Q8VE37
Entrez Gene ID: (Human) 1104, (Mouse) 100088
Molecular Function:
DNA binding protein
G-protein modulator
chromatin/chromatin-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.