Abnova
Manufacturer Code:PAB11748
Catalog # 89-115-352
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to N-terminus of human RCC1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. |
Rabbit polyclonal antibody raised against synthetic peptide of RCC1.
For Research Use Only
Protein Aliases: Cell cycle regulatory protein; CHC1; Chromosome condensation protein 1; guanine nucleotide-releasing protein; RCC1-I; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754
Entrez Gene ID: (Human) 1104
Molecular Function:
DNA binding protein
G-protein modulator
chromatin/chromatin-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.