Proteintech
Manufacturer Code:223351AP150UL
Catalog # 50-173-3833
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RCC1 Fusion Protein Ag17809 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cell cycle regulatory protein; Cell cycle regulatory protein CHC1 RCC1 RCC1 I; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: 4931417M11Rik; AI326872; CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754, (Mouse) Q8VE37
Entrez Gene ID: (Human) 1104, (Mouse) 100088, (Rat) 682908
Molecular Function: DNA binding protein G-protein modulator chromatin/chromatin-binding protein enzyme modulator guanyl-nucleotide exchange factor nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.