Abnova
Manufacturer Code:H00001104K
Catalog # 89-205-527
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Immunogen | A synthetic peptide of human RCC1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Rabbit monoclonal antibody raised against a human RCC1 peptide using ARM Technology.
For Research Use Only
Protein Aliases: Cell cycle regulatory protein; CHC1 RCC1-I; Chromosome condensation protein 1; guanine nucleotide-releasing protein; Regulator of chromosome condensation; SNHG3-RCC1 readthrough
Gene Aliases: CHC1; RCC1; RCC1-I; SNHG3-RCC1
UniProt ID: (Human) P18754
Entrez Gene ID: (Human) 1104
Molecular Function:
DNA binding protein
G-protein modulator
chromatin/chromatin-binding protein
enzyme modulator
guanyl-nucleotide exchange factor
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.