Invitrogen
Manufacturer Code:PA5102523
Catalog # PIPA5102523
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human EBAG9 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI835379 Cancer-associated surface antigen RCAS1 EB9 Ebag9 estrogen receptor binding site associated antigen 9 estrogen receptor-binding fragment-associated gene 9 Estrogen receptor-binding fragment-associated gene 9 protein HGNC:3123 PDAF Rcas1 Receptor-binding cancer antigen expressed on SiSo cells; Cancer-associated surface antigen RCAS1; Estrogen receptor-binding fragment-associated gene 9 protein; Receptor-binding cancer antigen expressed on SiSo cells
Gene Aliases: AI835379; EB9; EBAG9; PDAF; RCAS1
UniProt ID: (Human) O00559, (Mouse) Q9D0V7, (Rat) Q5PQP2
Entrez Gene ID: (Human) 9166, (Mouse) 55960, (Rat) 299864
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.