Proteintech
Manufacturer Code:129001AP150UL
Catalog # 50-172-8333
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | RCAN2 Fusion Protein Ag3597 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Calcineurin inhibitory protein ZAKI-4; Calcipressin 2 CSP2 DSCR1L1 hRCN2 MCIP2 RCAN2 regulator of calcineurin 2 ZAKI 4 ZAKI4; Calcipressin-2; Down syndrome candidate region 1-like 1; Down syndrome candidate region 1-like protein 1; Down syndrome critical region gene 1-like 1; MCIP2; myocyte-enriched calcineurin interactin protein 2; Myocyte-enriched calcineurin-interacting protein 2; Regulator of calcineurin 2; thyroid hormone-responsive (skin fibroblasts); Thyroid hormone-responsive protein ZAKI-4
Gene Aliases: CSP2; DSCR1L1; MCIP2; RCAN2; RCN2; ZAKI-4; ZAKI4
UniProt ID: (Human) Q14206, (Mouse) Q9JHG2
Entrez Gene ID: (Human) 10231, (Mouse) 53901
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.