Novus Biologicals
Manufacturer Code:NBP286769
Catalog # NB034391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human RCAN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Adapt78; ADAPT78 calcipressin-1 calcium and oxidant-inducible mRNA Down syndrome candidate region 1 Down syndrome critical region gene 1 Down syndrome critical region protein 1 DSC1 modulatory calcineurin-interacting protein 1 Myocyte-enriched calcineurin-interacting protein 1 near DSCR proline-rich protein regulator of calcineurin 1CSP1DSCR1MCIP1RCN1; Calcipressin-1; calcium and oxidant-inducible mRNA; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region protein 1; MCIP1; modulatory calcineurin-interacting protein 1; Myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; Regulator of calcineurin 1
Gene Aliases: ADAPT78; CSP1; DSC1; DSCR1; MCIP1; RCAN1; RCN1
UniProt ID: (Human) P53805
Entrez Gene ID: (Human) 1827
Molecular Function:
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.