Abnova
Manufacturer Code:PAB4386
Catalog # 89-131-396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human RCAN1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of RCAN1.
For Research Use Only
Protein Aliases: ADAPT78; Calcipressin-1; calcium and oxidant-inducible mRNA; CSP1; Down syndrome candidate region 1; Down syndrome critical region gene 1; Down syndrome critical region protein 1; DSC1; DSCR1; MCIP1; modulatory calcineurin-interacting protein 1; Myocyte-enriched calcineurin-interacting protein 1; near DSCR proline-rich protein; RCN1; Regulator of calcineurin 1
Gene Aliases: ADAPT78; CSP1; DSC1; DSCR1; MCIP1; RCAN1; RCN1
UniProt ID: (Human) P53805
Entrez Gene ID: (Human) 1827
Molecular Function: signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.