Invitrogen
Manufacturer Code:PA534519
Catalog # PIPA534519
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | An 18 amino acid peptide near the carboxy terminus of human RC3H1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: probable E3 ubiquitin-protein ligase Roquin; Protein Sanroque; RC3H1 Ring finger and CCCH-type domains 1 ring finger protein 198 RNF198 roquin; RING CCCH (C3H) domains 1; RING finger and C3H zinc finger protein 1; ring finger and CCCH-type domains 1; RING finger and CCCH-type zinc finger domain-containing protein 1; ring finger and CCCH-type zinc finger domains 1; RING finger protein 198; Roquin; Roquin-1
Gene Aliases: 5730557L09Rik; Gm551; KIAA2025; mKIAA2025; N28103; RC3H1; RGD1563512; RNF198; ROQUIN
UniProt ID: (Human) Q5TC82, (Mouse) Q69Z31
Entrez Gene ID: (Human) 149041, (Rat) 680586, (Mouse) 381305
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.