OriGene
Manufacturer Code:TA809421
Catalog # 50-168-2775
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4G10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 1-243 of human RBPJ produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AOS3 CBF1 IGKJRB IGKJRB1 KBF2 RBP-J RBPJK RBPSUH SUH csl; CBF-1; H-2K binding factor-2; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; RBP-J; RBP-J kappa; RBP-JK; Recombining binding protein suppressor of hairless; Renal carcinoma antigen NY-REN-30; suppressor of hairless homolog
Gene Aliases: AOS3; CBF1; csl; IGKJRB; IGKJRB1; KBF2; RBP-J; RBPJ; RBPJK; RBPSUH; SUH
UniProt ID: (Human) Q06330
Entrez Gene ID: (Human) 3516
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.