Invitrogen
Manufacturer Code:MA535438
Catalog # PIMA535438
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC0894 |
Immunogen | A synthesized peptide derived from human RBPJK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: AI843960 AOS3 BOS_6494 CBF1 CBF-1 csl IGKJRB Igkjrb1 Igkrsbp J kappa-recombination signal-binding protein KBF2 Rbpj RBP-J RBP-J kappa RBPJK RBPSUH Recombining binding protein suppressor of hairless Renal carcinoma antigen NY-REN-30 SUH; CBF-1; H-2K binding factor-2; immunoglobulin kappa J region recombination signal binding protein 1; J kappa-recombination signal-binding protein; RBP-J; RBP-J kappa; RBP-JK; Recombining binding protein suppressor of hairless; Renal carcinoma antigen NY-REN-30; suppressor of hairless homolog
Gene Aliases: AI843960; AOS3; CBF1; csl; IGKJRB; IGKJRB1; Igkrsbp; KBF2; RBP-J; RBP-J kappa; RBPJ; RBPJK; RBPSUH; SUH
UniProt ID: (Human) Q06330, (Mouse) P31266
Entrez Gene ID: (Human) 3516, (Mouse) 19664
Molecular Function:
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.